Learn More
Sino Biological Anti-VISTA/B7-H5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 203372T02100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human VISTA/VSIR fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-0552