Learn More
Sino Biological Human Vsir PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13482T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VISTA/GI24/B7-H5 (rh VISTA/GI24/B7-H5; Catalog#13482-H08H; AAH20568.1; Met1-Ala194). VISTA/GI24/B7-H5 specific IgG was purified by Human VISTA/GI24/B7-H5 affinity chromatography.
Catalog No. 50-237-4010