Learn More
Sino Biological Recombinant Human VISTA/B7-H5 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13482H08H1
Encompass_Preferred
A DNA sequence encoding the human Gi24 (AAH20568.1) (Met1-Ala194) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4234