Learn More
Sino Biological Recombinant Human VISTA/B7-H5 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 13482H49HB20
Encompass_Preferred
A DNA sequence encoding the human Vsir (AAH20568.1) (Met1-Ala194) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-2350