Learn More
Sino Biological Human VSNL1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14571T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VILIP-1 / VSNL1 (rh VILIP-1 / VSNL1; Catalog#14571-HNAE; Gly2-Lys191). VILIP-1 / VSNL1 specific IgG was purified by Human VILIP-1 / VSNL1 affinity chromatography.
Catalog No. 50-237-2131