Learn More
Sino Biological Anti-B7-H4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10738T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human B7-H4 (rh B7-H4; Catalog#10738-H08H; Q7Z7D3-1; Phe29-Ala258). B7-H4 specific IgG was purified by Human B7-H4 affinity chromatography.
Catalog No. 50-241-1231