Learn More
Sino Biological Recombinant Human B7-H4 Protein (mFc Tag), HPLC-verified
Supplier:  Sino Biological 10738H05H200
Encompass_Preferred
A DNA sequence encoding the human VTCN1 (NP_078902.2) (Phe29-Ala258) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-255-5187