Learn More
Sino Biological Recombinant Human B7-H4 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10738H08HB20
Encompass_Preferred
A DNA sequence encoding the human VTCN1(Q7Z7D3-1) (Phe29-Ala258) with a C-terminal polyhistidine tag was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5184