Learn More
Sino Biological Recombinant Human B7-H4 Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10738H02HB20
Encompass_Preferred
A DNA sequence encoding the human VTCN1 (Q7Z7D3-1) (Phe29-Ala258) was expressed, fused with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5188