Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VTN (multimeric) native Protein from human plasma

Catalog No. 89967190 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-190 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-190 Supplier Abnova™ Supplier No. P4954
Add to Cart
Add to Cart

Used for SDS-PAGE

The protein encoded by this gene is a member of the pexin family. It is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. It is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond. [provided by RefSeq]

Specifications

Concentration 3.4 mg/mL
For Use With (Application) SDS-PAGE
Formulation In 0.05M sodium phosphate, 0.1M NaCl, pH 7.4
Gene ID (Entrez) 7448
Molecular Weight (g/mol) 75 monomer
Name VTN (Human) Native Protein
Preparation Method Native protein purified from human plasma
Purification Method Non-denaturing chromatography
Quantity 100 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias V75/VN/VNT
Common Name VTN
Gene Symbol VTN
Species Human
Protein Tag None
Expression System Native protein purified from human plasma
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.