Learn More
Sino Biological Recombinant Human Von Willebrand Factor/vWF Protein (A1 Domain, His Tag)
Catalog No. 502992689
Encompass_Preferred
A DNA sequence encoding the Human vWF (P04275) (Asp1277-Ile1453) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2689 Supplier Sino Biological Supplier No. 10973H08E1
A DNA sequence encoding the Human vWF (P04275) (Asp1277-Ile1453) was expressed with a polyhistidine tag at the C-terminus.