Learn More
Sino Biological Anti-Von Willebrand Factor/vWF Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10973MM0520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VWF / Von Willebrand Factor (rh VWF / Von Willebrand Factor; Catalog#10973-H08C; AAB59458.1; Met1-Lys2813). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0357