Learn More
Haematologic Technologies Inc HUMAN VWF (VIII FREE) 100UG
Supplier:  Haematologic Technologies Inc HCVWF0191
Encompass_Preferred
Coagulation protein, Human von Willebrand Factor (VIII Free), 100 micrograms, Purified from platelet plasma, supplied in 25mM Citrate, 100mM NaCl, 100mM Glycine, pH 6.8, store at -80 Celsius.
Catalog No. 50-883-487