Learn More
Sigma Aldrich Fine Chemicals Biosciences Human VWf von Willebrand
Catalog No. 502906533
Encompass
Human VWf/von Willebrand factor ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro assay for the quantitative measurement of a target protein in biological samples such as serum plasma cell culture supernatants urine and cell and tissue lysates.
Catalog No. 50-290-6533 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RAB05561KT
Human VWf/von Willebrand factor ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro assay for the quantitative measurement of a target protein in biological samples such as serum plasma cell culture supernatants urine and cell and tissue lysates.