Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WBP1 Full-length ORF (NP_036609.1, 1 a.a. - 269 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967229 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-229 25 μg
89-967-228 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-229 Supplier Abnova™ Supplier No. H00023559P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The globular WW domain is composed of 38 to 40 semiconserved amino acids shared by proteins of diverse functions including structural, regulatory, and signaling proteins. The domain is involved in mediating protein-protein interactions through the binding of polyproline ligands. This gene encodes a WW domain binding protein, which binds to the WW domain of Yes kinase-associated protein by a conserved region: XPPXY motif. The function of this protein has not been determined. [provided by RefSeq]

Sequence: MARASSGNGSEEAWGALRAPQQQLRELCPGVNNQPYLCESGHCCGETGCCTYYYELWWFWLLWTVLILFSCCCAFRHRRAKLRLQQQQRQREINLLAYHGACHGAGPFPTGSLLDLRFLSTFKPPAYEDVVHRPGTPPPPYTVAPGRPLTASSEQTCCSSSSSCPAHFEGTNVEGVSSHQSAPPHQEGEPGAGVTPASTPPSCRYRRLTGDSGIELCPCPASGEGEPVKEVRVSATLPDLEDYSPCALPPESVPQIFPMGLSSSEGDIP

Specifications

Accession Number NP_036609.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23559
Molecular Weight (g/mol) 55.5kDa
Name WBP1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MARASSGNGSEEAWGALRAPQQQLRELCPGVNNQPYLCESGHCCGETGCCTYYYELWWFWLLWTVLILFSCCCAFRHRRAKLRLQQQQRQREINLLAYHGACHGAGPFPTGSLLDLRFLSTFKPPAYEDVVHRPGTPPPPYTVAPGRPLTASSEQTCCSSSSSCPAHFEGTNVEGVSSHQSAPPHQEGEPGAGVTPASTPPSCRYRRLTGDSGIELCPCPASGEGEPVKEVRVSATLPDLEDYSPCALPPESVPQIFPMGLSSSEGDIP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC15305/WBP-1
Common Name WBP1
Gene Symbol WBP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.