Learn More
Techcyte Human Wet Mount O&P Fecal Analysis
Description
- Software that facilitates the finding, counting, and pre-classification of human fecal parasites from a digital image
- Scanned image is of a designated portion of a wet mount iodine stained fecal smear from a coverslipped glass slide
- Device is intended for screening of human fecal samples for in-vitro diagnostic use by trained technologists
- Software locates, counts, and pre-classifies parasite cysts, parasite trophozoites, red blood cells, white blood cells, and yeast from a digital image produced by a high-definition digital slide scanner
- Slide needs to be a iodine-stained, monolayer-prepared sample
Specifications
Specifications
| Product Type | Detection Software |
| For Use With (Application) | Analyzing Data |
| Operating System | Microsoft Windows 10 IoT 64-bit |
For investigational use only in the US.