Learn More
Sino Biological Anti-HE4/WFDC2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12609MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HE4/WFDC2/WAP5 (rh HE4/WFDC2/WAP5; Catalog#12609-H08H; NP_006094.3; Met1-Phe124). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4362