Learn More
Sino Biological Recombinant Anti-HE4/WFDC2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12609R404100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human WFDC2 / HE4 (rh WFDC2 / HE4; Catalog#12609-H08H; NP_006094.3; Met1-Phe124).
Catalog No. 50-255-4355