Learn More
Amsbio LLC Human Whole Blood|  up to 10ml
Catalog No. 502563596
Encompass
Human Whole Blood up to 10ml. Anticoagulant: ACD or EDTA are Standard--specify if another is needed. *Draw volume may vary according to anticoagulant*. Expiration: 21 days after collection when stored at 2-8C (Shipped after pathogen testing is complete)
Catalog No. 50-256-3596 Supplier Amsbio LLC Supplier No. SERWB10ML
Human Whole Blood up to 10ml. Anticoagulant: ACD or EDTA are Standard--specify if another is needed. *Draw volume may vary according to anticoagulant*. Expiration: 21 days after collection when stored at 2-8C (Shipped after pathogen testing is complete)