Learn More
Biomatik Corporation Human Wilms Tumor Protein (WT1) CLIA Kit
Supplier:  Biomatik Corporation EKU09488
Encompass_Preferred
Human Wilms Tumor Protein (WT1) CLIA Kit for detecting WT1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P19544. Gene ID: 7490. Target Synonyms: WAGR; GUD; WIT-2; WT33. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7454