Learn More
Biomatik Corporation Human Wingless Type MMTV Integration Site Family, Member 3 (WNT3) CLIA Kit
Supplier:  Biomatik Corporation EKU09986
Encompass_Preferred
Human Wingless Type MMTV Integration Site Family, Member 3 (WNT3) CLIA Kit for detecting WNT3 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 15.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P56703. Gene ID: 7473. Target Synonyms: INT4; Proto-oncogene Int-4 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6674