Learn More
Abcam Human WISP-1 ELISA Kit (CCN4), 1 x 96 Tests
Supplier:  Abcam AB1554451X96TESTS
Encompass_Preferred
Human WISP-1 ELISA Kit (CCN4) is a Sandwich (quantitative) ELISA kit for the measurement of Human WISP-1 (CCN4) in Human in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2255087