Learn More
Gibco™ Human WISP-1 Recombinant Protein, PeproTech®
Description
120-18-1MG will be provided as 2 x 500 μg (120-18-500UG). Recombinant Human WISP-1 is a 38.1 kDa protein containing 345 amino acid residues. It is composed of four distinct structural domains (modules): the IGF binding protein (IGFBP) domain, the von Willebrand Factor C (VWFC) domain, the thrombospondin type-1 repeat (TSP type-1) domain, and a C-terminal cysteine knot-like (CTCK) domain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | O95388 |
| For Use With (Application) | Bioactivity, Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 8840 |
| Molecular Weight (g/mol) | 38.1 kDa |
| Name | Human WISP-1 |
| Purification Method | Purified |
| Quantity | 5 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.