Learn More
BIOHIPPO Human WISP3 Microsample
SDP
Catalog No. 5000313939
Encompass
Human WISP3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of WISP3 in Serum plasma and other biological fluids It provides a detection range of 93 75-6000 pg/mL with a sensitivity of 36 14 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human WISP3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human WISP3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human WISP3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13939 Supplier BIOHIPPO Supplier No. BHE15210483
Human WISP3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of WISP3 in Serum plasma and other biological fluids It provides a detection range of 93 75-6000 pg/mL with a sensitivity of 36 14 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human WISP3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human WISP3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human WISP3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color