Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Wnt-1 Recombinant Protein, PeproTech®

Catalog No. 120172UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-17-2UG 2 μg
12017100UG 100 μg
120-17-10UG 10 μg
120-17-1MG 1 mg
12017250UG 250 μg
12017500UG 500 μg
120-17-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 120-17-2UG Supplier Gibco™ Supplier No. 120172UG
  • $92.50
    $78.65 / Each of 1

    Save $13.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Wnt-1 is a 38.4 kDa, non-glycosylated protein containing 343 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Wnt proteins constitute a family of secreted glyocoproteins that activate signal transduction pathways to mediate tissue homeostasis, cell fate, cell proliferation, and self-renewal. The canonical signaling pathway is initiated upon Wnt binding to a Frizzled family receptor and an LRP5/LRP6 coreceptor, which leads to activation of the beta-catenin cascade. Non-canonical Wnt signals are activated by binding to a Frizzled family receptor and ROR2 coreceptor. Extensive research has shown that deregulation of these pathways or its components can lead to cancer. Human Wnt-1 is encoded as a 370 amino acid precursor that, upon secretion, is cleaved into a 343 amino acid (42 kDa) mature form. At the amino acid level, mature human Wnt-1 is approximately 99% identical to mature mouse Wnt-1. Depending on the tissue, Wnt-1 can activate the canonical and non-canonical signaling pathways. Studies have demonstrated its involvement in a variety of functions, including embryonic development and carcinogenesis. Wnt-1 signaling can also induce the phosphorylation and subsequent degradation of c-Myb.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04628
For Use With (Application) Bioactivity, Control, Western Blot Control
Formulation Protein with no preservative
Gene ID (Entrez) 7471
Molecular Weight (g/mol) 38.4 kDa
Name Human Wnt-1
Purification Method Purified
Quantity 2 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias BMND16; INT1; Int-1; murine mammary tumor virus integration site; OI15; Proto-oncogene Int-1; proto-oncogene Int-1 homolog; proto-oncogene protein Wnt-1; proto-oncogene Wnt-1; sw; swaying; wingless-related MMTV integration site 1; Wingless-type MMTV integration site 1 homolog; Wingless-type MMTV integration site 1, homolog; wingless-type MMTV integration site family member 1; wingless-type MMTV integration site family, member 1; wingless-type MMTV integration site family, member 1 (oncogene INT1); Wnt family member 1; Wnt1; Wnt-1
Common Name WNT1
Gene Symbol WNT1
Biological Activity The ED50 was determined by its ability to enhance BMP-2 induced alkaline phosphatase production by murine ATDC5 cells. The expected ED50 for this effect is 1.5 - 2.5 ng/ml in the presence of 200 ng/ml of human BMP-2.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence ANSSGRWWGI VNVASSTNLL TDSKSLQLVL EPSLQLLSRK QRRLIRQNPG ILHSVSGGLQ SAVRECKWQF RNRRWNCPTA PGPHLFGKIV NRGCRETAFI FAITSAGVTH SVARSCSEGS IESCTCDYRR RGPGGPDWHW GGCSDNIDFG RLFGREFVDS GEKGRDLRFL MNLHNNEAGR TTVFSEMRQE CKCHGMSGSC TVRTCWMRLP TLRAVGDVLR DRFDGASRVL YGNRGSNRAS RAELLRLEPE DPAHKPPSPH DLVYFEKSPN FCTYSGRLGT AGTAGRACNS SSPALDGCEL LCCGRGHRTR TQRVTERCNC TFHWCCHVSC RNCTHTRVLH ECL
Content And Storage -20°C
Activity The ED50 was determined by its ability to enhance BMP-2 induced alkaline phosphatase production by murine ATDC5 cells. The expected ED50 for this effect is 1.5 - 2.5 ng/ml in the presence of 200 ng/ml of human BMP-2.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.