Learn More
Raybiotech Inc Human Wnt-4 ELISA Kit
Supplier:  Raybiotech Inc ELHWNT41
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 1.22 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Wnt-4. Other species not determined.
Catalog No. 50-194-9860