Learn More
Raybiotech Inc Human Wnt-9a ELISA Kit
Supplier:  Raybiotech Inc ELHWNT9A1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.75 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Wnt-9a.
Catalog No. 50-194-9861