Learn More
Gibco™ Human Wnt-9b Recombinant Protein, PeproTech®
Description
120-49-1MG will be provided as 10 x 100 μg (120-49-100UG). 120-49-250UG will be provided as multiple 50 μg (120-49-50UG) and 100 μg (120-49-100UG) vials. 120-49-500UG will be provided as 5 x 100 μg (120-49-100UG). Recombinant Human Wnt-9b is a monomeric glycoprotein containing 335 amino acid residues and has a calculated molecular weight of 36.9 kDa. Due to glycosylation, Recombinant Human Wnt-9b migrates at an apparent molecular weight of approximately 49-54 kDa by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | O14905 |
| For Use With (Application) | Bioactivity, Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 7484 |
| Molecular Weight (g/mol) | 36.9 kDa |
| Name | Human Wnt-9b |
| Purification Method | Purified |
| Quantity | 20 μg |
| Source | CHO cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.