Learn More
Biomatik Corporation Human Wolfram Syndrome Protein 1 (WFS1) CLIA Kit
Supplier:  Biomatik Corporation EKU09982
Encompass_Preferred
Human Wolfram Syndrome Protein 1 (WFS1) CLIA Kit for detecting WFS1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O76024. Gene ID: 7466. Target Synonyms: DFNA14; DFNA38; DFNA6; DIDMOAD; WFRS; WFS; Wolframin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6420