Learn More
Sino Biological Anti-WTAP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12018RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human WTAP (rh WTAP; Catalog#12018-H09E; Q15007-2; Met1-Arg151). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1487