Learn More
Biomatik Corporation Human WW Domain Containing E3 Ubiquitin Protein Ligase 2 (WWP2) ELISA Kit
Supplier:  Biomatik Corporation EKU08230
Encompass_Preferred
Human WW Domain Containing E3 Ubiquitin Protein Ligase 2 (WWP2) ELISA Kit for detecting WWP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00308. Gene ID: 11060. Target Synonyms: AIP2; WWp2-like; Atrophin-1-interacting protein 2; NEDD4-like E3 ubiquitin-protein ligase WWP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6305