Learn More
Sino Biological Human WWP2 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 13125MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human WWP2 (rh WWP2; Catalog#13125-H20B; O00308; Met1-Glu870). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2919