Learn More
Biomatik Corporation Human X-Box Binding Protein 1 (XBP1) CLIA Kit
Supplier:  Biomatik Corporation EKU10003
Encompass_Preferred
Human X-Box Binding Protein 1 (XBP1) CLIA Kit for detecting XBP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P17861. Gene ID: 7494. Target Synonyms: TREB5; XBP2; Tax-responsive element-binding protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8384