Learn More
Sino Biological Anti-Lymphotactin/XCL1/SCYC1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10672T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human XCL1 (rh XCL1; Catalog#10672-H08B; P47992; Met1-Gly114). XCL1 specific IgG was purified by Human XCL1 affinity chromatography.
Catalog No. 50-240-9226