Learn More
Sino Biological Recombinant Human Lymphotactin/XCL1/SCYC1 Protein (His Tag)
Supplier:  Sino Biological 10672H08B1
Encompass_Preferred
A DNA sequence encoding the human XCL1 (P47992) (Met 1-Gly 114) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5228