Learn More
Biomatik Corporation Human Xeroderma Pigmentosum, Complementation Group G (XPG) CLIA Kit
Supplier:  Biomatik Corporation EKU10002
Encompass_Preferred
Human Xeroderma Pigmentosum, Complementation Group G (XPG) CLIA Kit for detecting XPG in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P28715. Gene ID: 2073. Target Synonyms: ERCM2; ERCC5; UVDR; XPGC; Excision Repair Cross-Complementing Rodent Repair Deficiency 5; Cockayne Syndrome; DNA repair protein complementing XP-G cells. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7450