Learn More
Biomatik Corporation Human Xeroderma Pigmentosum, Complementation Group C (XPC) CLIA Kit
Supplier:  Biomatik Corporation EKU10001
Encompass_Preferred
Human Xeroderma Pigmentosum, Complementation Group C (XPC) CLIA Kit for detecting XPC in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q01831. Gene ID: 7508. Target Synonyms: XP-C; XP3; XPCC; RAD4; Xeroderma Pigmentosum Group C Protein; DNA repair protein complementing XP-C cells. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7643