Learn More
Biomatik Corporation Human Xeroderma Pigmentosum, Complementation Group G (XPG) ELISA Kit
Supplier:  Biomatik Corporation EKU08240
Encompass_Preferred
Human Xeroderma Pigmentosum, Complementation Group G (XPG) ELISA Kit for detecting XPG in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.128ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28715. Gene ID: 2073. Target Synonyms: ERCM2; ERCC5; UVDR; XPGC; Excision Repair Cross-Complementing Rodent Repair Deficiency 5; Cockayne Syndrome; DNA repair protein complementing XP-G cells. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8688