Learn More
Biomatik Corporation Human Xeroderma Pigmentosum, Complementation Group D (XPD) ELISA Kit
Supplier:  Biomatik Corporation EKU09100
Encompass_Preferred
Human Xeroderma Pigmentosum, Complementation Group D (XPD) ELISA Kit for detecting XPD in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18074. Gene ID: 2068. Target Synonyms: CXPD; BTF2 p80; EM9; TTD; ERCC2; TFIIH basal transcription factor complex 80 kDa; De Sanctis-Cacchione; Excision Repair Cross-Complementing Rodent Repair Deficiency 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6190