Learn More
Biomatik Corporation Human X-linked Inhibitor Of Apoptosis Protein (XIAP) CLIA Kit
Supplier:  Biomatik Corporation EKU10005
Encompass_Preferred
Human X-linked Inhibitor Of Apoptosis Protein (XIAP) CLIA Kit for detecting XIAP in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P98170. Gene ID: 331. Target Synonyms: BIRC4; API3; ILP1; MIHA; XLP2; Baculoviral IAP Repeat-Containing 4; E3 ubiquitin-protein ligase XIAP; IAP-like; Inhibitor of apoptosis protein 3; Inhibitor Of Apoptosis Protein, X-Linked. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5606