Learn More
Sino Biological Recombinant Human XPNPEP2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11903H08H50
Encompass_Preferred
A DNA sequence encoding the human XPNPEP2 (O43895) (Met 1-Ala 650) without the pro peptide was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7289