Learn More
Abcam Human YKL-40 ELISA Kit, 1 x 96 Tests
Catalog No. 502656945
Encompass_Preferred
Human YKL-40 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human YKL-40 with a sensitivity of 3.9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF) - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6945 Supplier Abcam Supplier No. AB2557191X96TESTS
Add to Cart
Add to Cart
Human YKL-40 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human YKL-40 with a sensitivity of 3.9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF) - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement