Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human YME1L1 Partial ORF (NP_647474.1, 514 a.a. - 612 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967611 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-611 25 μg
89-967-610 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-611 Supplier Abnova™ Supplier No. H00010730Q01L

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: EILKWYLNKIKFDQSVDPEIIARGTVGFSGAELENLVNQAALKAAVDGKEMVTMKELEFSKDKILMGPERRSVEIDNKNKTITAYHESGHAIIAYYTKD

Specifications

Accession Number NP_647474.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10730
Molecular Weight (g/mol) 36.63kDa
Name YME1L1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen EILKWYLNKIKFDQSVDPEIIARGTVGFSGAELENLVNQAALKAAVDGKEMVTMKELEFSKDKILMGPERRSVEIDNKNKTITAYHESGHAIIAYYTKD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FTSH/MEG4/PAMP/YME1L
Common Name YME1L1
Gene Symbol YME1L1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.