Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human YOD1 (aa 116-191) Control Fragment Recombinant Protein

Catalog No. RP94482
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP94482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP94482 Supplier Invitrogen™ Supplier No. RP94482
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55837. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Deubiquitinating enzymes (DUBs) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA8 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5VVQ6
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 55432
Name Human YOD1 (aa 116-191) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 9930028C20Rik; DUBA8; DUBA-8; fe01a10; HIN7; HIN-7; HIV-1-induced protease 7; HIV-induced protein-7-like protease; hsHIN7; hypothetical protein LOC550411; OTU domain containing 2; OTU domain-containing protein 2; OTUD2; PRO0907; Ubiquitin thioesterase OTU1; wu:fe01a10; Yod1; YOD1 deubiquitinase; YOD1 OTU deubiquinating enzyme 1 homolog; YOD1 OTU deubiquinating enzyme 1 homolog ( yeast); YOD1 OTU deubiquitinating enzyme 1 homologue; YOD1 OTU deubiquitinating enzyme 1 homologue (S. cerevisiae); zgc:112182
Common Name YOD1
Gene Symbol YOD1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DMLIIEEDQTRPRSSPAFTKRGASSYVRETLPVLTRTVVPADNSCLFTSVYYVVEGGVLNPACAPEMRRLIAQIVA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.