Learn More
Sino Biological Human YWHAE PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10842T54100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human 14-3-3 epsilon/YWHAE (rh 14-3-3 epsilon/YWHAE; Catalog#10842-HNCE; NP_006752.1; Met1-Gln255). 14-3-3 epsilon/YWHAE specific IgG was purified by Human 14-3-3 epsilon/YWHAE affinity chromatography.
Catalog No. 50-237-3113