Learn More
Sino Biological Anti-14-3-3 eta/YWHAH Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107196T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human 14-3-3 eta/YWHAH, and purified by antigen affinity chromatography.
Catalog No. 50-241-0428