Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human YY1 ELISA Kit

Catalog No. EH488RBX10
Encompass_Preferred
Change view
Click to view available options
10 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EH488RBX10 10 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH488RBX10 Supplier Invitrogen™ Supplier No. EH488RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human YY1 quantitates human YY1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human YY1.

YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P25490
Assay Range 0.8-200 ng/mL
Assay Sensitivity 0.8 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AW488674; DELTA; delta transcription factor; fb59g10; INO80 complex subunit S; INO80S; NF-E1; NMP1; NMP-1; transcription factor Yy1; transcriptional repressor protein YY1; transcriptional repressor protein YY1a; Ucrbp; UCRBP transcription factor; UCR-motif DNA-binding protein; wu:fa16g07; wu:fb59g10; Yin and yang 1; Yin and Yang 1 protein; Yin Yang 1; YIN-YANG-1; yy1; YY-1; yy1 protein; YY1 transcription factor; YY1 transcription factor a; yy1aAW488674; DELTA; delta transcription factor; fb59g10; INO80 complex subunit S; INO80S; NF-E1; NMP1; NMP-1; transcription factor Yy1; transcriptional repressor protein YY1; transcriptional repressor protein YY1a; Ucrbp; UCRBP transcription factor; UCR-motif DNA-binding protein; wu:fa16g07; wu:fb59g10; Yin and yang 1; Yin and Yang 1 protein; Yin Yang 1; YIN-YANG-1; yy1; YY-1; yy1 protein; YY1 transcription factor; YY1 transcription factor a; yy1a;
Gene ID (Entrez) 7528
Gene Symbol YY1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target YY1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.