Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ZBTB40 Synthetic Peptide

Catalog No. PEP1442 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1442 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1442 Supplier Invitrogen™ Supplier No. PEP1442
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 19 amino acid peptide near the amino terminus of human ZBTB40. PEP-1442 can be used as a blocking peptide with polyclonal antibody PA5-34400.

May be involved in transcriptional regulation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9923
Species Human
Purification Method purified
Accession Number Q9NUA8
Common Name ZBTB40
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias BC059177; C230087D24; Gm571; KIAA0478; mKIAA0478; RGD1309866; ZBTB40; zinc finger and BTB domain containing 40; zinc finger and BTB domain-containing protein 40; ZNF923
Gene Symbol ZBTB40
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.