Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ZCCHC4 Control Fragment Recombinant Protein

Catalog No. RP102673
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP102673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP102673 Supplier Invitrogen™ Supplier No. RP102673
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57319. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The ZCCHC4 gene encodes zinc finger CCHC domain-containing protein 4, a member of the zinc finger CCHC-type (ZCCHC) superfamily characterized by a consensus sequence C-X2-C-X4-H-X4-C. This gene plays a vital role in binding single-stranded nucleic acids, particularly RNAs, and is involved in multiple cellular processes. Notably, ZCCHC4 has been implicated in cancer, with aberrantly high expression linked to poor prognosis and chemoresistance in various cancers, including hepatocellular carcinoma, pancreatic cancer, colon cancer, and esophageal cancer. The protein exhibits RNA-modifying capabilities, influencing cell growth and survival. Recent studies have identified the structural mechanisms through which ZCCHC4 interacts with RNA substrates, offering insights into its role in regulating cancer progression and therapeutic resistance. In esophageal cancer, downregulation of ZCCHC4 may enhance sensitivity to chemotherapeutic agents like cisplatin, indicating its potential as a target for cancer treatment strategies.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H5U6
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 29063
Name Human ZCCHC4 Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 4930449I23Rik; DHHC domain-containing zinc finger protein; HSPC052; rRNA N6-adenosine-methyltransferase ZCCHC4; Zcchc4; ZGRF4; zinc finger CCHC domain-containing protein 4; zinc finger CCHC-type containing 4; zinc finger, CCHC domain containing 4; zinc finger, GRF-type containing 4
Common Name ZCCHC4
Gene Symbol ZCCHC4
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FESRICQFFPSFQMLDYQVDYDNHALYKHGKTGRKQSPVRIFTNIPPNKIILPTEEGYRFCSPCQRYVSLENQHCEHCNSCTSKDGRKW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.