Learn More
Sino Biological Anti-ZDHHC21 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 208048T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human ZDHHC21 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-299-2803